Thermo Scientific Chemicals

alpha-Fluorophenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: L01745.MC
100 mg, Each
Thermo Scientific Chemicals

alpha-Fluorophenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: L01745.MC
100 mg, Each
Quantity
Catalog number: L01745.MC
also known as L01745-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1578-63-8
IUPAC Name
(2S)-2-fluoro-2-phenylacetate
Molecular Formula
C8H6FO2
InChI Key
ATPPNMLQNZHDOG-ZETCQYMHSA-M
SMILES
[O-]C(=O)[C@@H](F)C1=CC=CC=C1
Form
Crystalline powder or crystals
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Melting Point (clear melt)
77-86?C
Appearance (Color)
White to pale cream
Assay (Silylated GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text