Thermo Scientific Chemicals

3-Methyl-2-furoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L01763.06
5 g, Each
Thermo Scientific Chemicals

3-Methyl-2-furoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L01763.06
5 g, Each
Quantity
Catalog number: L01763.06
also known as L01763-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4412-96-8
IUPAC Name
3-methylfuran-2-carboxylic acid
Molecular Formula
C6H6O3
InChI Key
BNYIQEFWGSXIKQ-UHFFFAOYSA-N
SMILES
CC1=C(OC=C1)C(O)=O
Form
Powder or granules
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (Silylated GC)
≥98.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text