2,4-Pentanediol was used in the synthesis of chelated multinuclear complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Pentanediol was used in the synthesis of chelated multinuclear complexes.
Solubility
Fully miscible.
Notes
Hygroscopic. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
J. G. Pritchard.; R. L. Vollmer. The meso and Racemic Forms of 2,4-Pentanediol and Certain of Their Derivatives. J. Org. Chem. 1963, 28, (6), 1545-1549.
Takashi Sugimura.; Tohru Futagawa.; Akira Tai. Diastereo-differentiating simmons-smith reaction using 2,4-pentanediol as a chiral auxiliary. Tetrahedron Letters. 1988, 29, (45), 5775-5778.