Thermo Scientific Chemicals

9-Fluorenol, 98+%, Thermo Scientific Chemicals

Catalog number: L01917.14
25 g, Each
Thermo Scientific Chemicals

9-Fluorenol, 98+%, Thermo Scientific Chemicals

Catalog number: L01917.14
25 g, Each
Quantity
Catalog number: L01917.14
also known as L01917-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1689-64-1
IUPAC Name
9H-fluoren-9-ol
Molecular Formula
C13H10O
InChI Key
AFMVESZOYKHDBJ-UHFFFAOYSA-N
SMILES
OC1C2=CC=CC=C2C2=CC=CC=C12
Appearance (Color)
White to pale cream or pale yellow
Assay (GC)
≥98.0% (non-U.S. specification)
Identification (FTIR)
Conforms (non-U.S. specification)
Assay from Suppliers CofA
≥98.0% (U.S. specification)
Form
Crystals or powder or crystalline powder or needles or granules

Description

9-Fluorenol is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Fluorenol is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Insoluble in water.

Notes
Protect from moisture, heat. Store in cool place. Keep container tightly
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text