Thermo Scientific Chemicals

5,6-Dihydrouracil, 97%, Thermo Scientific Chemicals

Catalog number: L01918.06
5 g, Each
Thermo Scientific Chemicals

5,6-Dihydrouracil, 97%, Thermo Scientific Chemicals

Catalog number: L01918.06
5 g, Each
Quantity
Catalog number: L01918.06
also known as L01918-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
504-07-4
IUPAC Name
1,3-diazinane-2,4-dione
Molecular Formula
C4H6N2O2
InChI Key
OIVLITBTBDPEFK-UHFFFAOYSA-N
SMILES
O=C1CCNC(=O)N1
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder or lumps
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms

Description

5,6-Dihydrouracil acts as an intermediate in the catabolism of uracil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Dihydrouracil acts as an intermediate in the catabolism of uracil.

Solubility
Soluble in Sodium Hydroxide.

Notes
Incompatible with oxidizing agents.Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text