Thermo Scientific Chemicals

Phenanthrene, 98%, Thermo Scientific Chemicals

Catalog number: L01921.30
250 g, Each
Thermo Scientific Chemicals

Phenanthrene, 98%, Thermo Scientific Chemicals

Catalog number: L01921.30
250 g, Each
Quantity
Catalog number: L01921.30
also known as L01921-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85-01-8
IUPAC Name
phenanthrene
Molecular Formula
C14H10
InChI Key
YNPNZTXNASCQKK-UHFFFAOYSA-N
SMILES
C1=CC=C2C(C=CC3=CC=CC=C23)=C1
Appearance (Color)
White to cream to pale brown or grey
Form
Crystals or powder or crystalline powder or flakes
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
97-101°C

Description

Phenanthrene is generally used for dye production, synthetic resin, preservative. It is also used in agrochemical and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenanthrene is generally used for dye production, synthetic resin, preservative. It is also used in agrochemical and pharmaceutical intermediates.

Solubility
Soluble in Ether, Benzene, Chloroform, Toluene. Insoluble in water.

Notes
Light sensitive. Handle and store under inert gas. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text