Thermo Scientific Chemicals

3-Thiophenemethanol, 97%, Thermo Scientific Chemicals

Catalog number: L01935.06
5 g, Each
Thermo Scientific Chemicals

3-Thiophenemethanol, 97%, Thermo Scientific Chemicals

Catalog number: L01935.06
5 g, Each
Quantity
Catalog number: L01935.06
also known as L01935-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71637-34-8
IUPAC Name
(thiophen-3-yl)methanol
Molecular Formula
C5H6OS
InChI Key
BOWIFWCBNWWZOG-UHFFFAOYSA-N
SMILES
OCC1=CSC=C1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥96.0%

Description

3-Thiophenemethanol was used in the preparation of 3-substituted thiophene conducting copolymers which has potential applications in electrochromic displays.1 It was also used in the synthesis of 4-(thiophene-3-ylmethoxy)phthalonitrile

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Thiophenemethanol was used in the preparation of 3-substituted thiophene conducting copolymers which has potential applications in electrochromic displays.1 It was also used in the synthesis of 4-(thiophene-3-ylmethoxy)phthalonitrile

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text