2,4,6-Trimethylbenzyl alcohol, 98+%, Thermo Scientific Chemicals
2,4,6-Trimethylbenzyl alcohol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,6-Trimethylbenzyl alcohol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01952.06
also known as L01952-06
Price (USD)/ Each
66.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
66.00
Add to cart
2,4,6-Trimethylbenzyl alcohol, 98+%, Thermo Scientific Chemicals
Catalog numberL01952.06
Price (USD)/ Each
66.00
-
Add to cart
Chemical Identifiers
CAS4170-90-5
IUPAC Name(2,4,6-trimethylphenyl)methanol
Molecular FormulaC10H14O
InChI KeyLODDFDHPSIYCTK-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(CO)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Melting Point (clear melt)84.5-90.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only