Thermo Scientific Chemicals

Cyclopentylamine, 99%, Thermo Scientific Chemicals

Catalog number: L01966.14
25 g, Each
Thermo Scientific Chemicals

Cyclopentylamine, 99%, Thermo Scientific Chemicals

Catalog number: L01966.14
25 g, Each
Quantity
Catalog number: L01966.14
also known as L01966-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1003-03-8
IUPAC Name
cyclopentanamine
Molecular Formula
C5H11N
InChI Key
NISGSNTVMOOSJQ-UHFFFAOYSA-N
SMILES
NC1CCCC1
Identification (FTIR)
Conforms
Refractive Index
1.4480-1.4510 @20?C
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%

Description

Cyclopentylamine acts as an important raw material and intermediate used in organic synthesis and pharmaceuticals. Further, it serves as chemokine receptor 2 antagonists. It acts as a ligand in coordination chemistry and forms cis-dichlorobis(cyclopentylamine)platinum(II) complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentylamine acts as an important raw material and intermediate used in organic synthesis and pharmaceuticals. Further, it serves as chemokine receptor 2 antagonists. It acts as a ligand in coordination chemistry and forms cis-dichlorobis(cyclopentylamine)platinum(II) complex.

Solubility
Miscible with water, acetone and benzene.

Notes
Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text