Thermo Scientific Chemicals

Phenoxazine, 98%, Thermo Scientific Chemicals

Catalog number: L01969.03
1 g, Each
Thermo Scientific Chemicals

Phenoxazine, 98%, Thermo Scientific Chemicals

Catalog number: L01969.03
1 g, Each
Quantity
Catalog number: L01969.03
also known as L01969-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
135-67-1
IUPAC Name
10H-phenoxazine
Molecular Formula
C12H9NO
InChI Key
TZMSYXZUNZXBOL-UHFFFAOYSA-N
SMILES
N1C2=CC=CC=C2OC2=CC=CC=C12
Appearance (Color)
Cream to brown or grey or grey/brown
Assay (GC)
≥97.5%
Assay (HPLC)
≥97.5%
Infrared spectrum
Conforms
Melting Point (clear melt)
152-159?C

Description

Phenoxazine is used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenoxazine is used as primary and secondary intermediates.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text