Phenoxazine is used as primary and secondary intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenoxazine is used as primary and secondary intermediates.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
RUO – Research Use Only
Thimmaiah KN; Horton JK; Qian XD; Beck WT; Houghton JA; Houghton PJ. Structural determinants of phenoxazine type compounds required to modulate the accumulation of vinblastine and vincristine in multidrug-resistant cell lines.Cancer Communications.1990, 2 (7), 249-259.
W. Michael Flanagan; Richard W. Wagner; Deborah Grant; Kuei-Ying Lin1 & Mark D. Matteucci. Cellular penetration and antisense activity by a phenoxazine-substituted heptanucleotide.Nature Biotechnology.1999, 17 48 - 52.