5,6-Dihydro-5-methyluracil, 98+%, Thermo Scientific Chemicals
5,6-Dihydro-5-methyluracil, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5,6-Dihydro-5-methyluracil, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01996.06
also known as L01996-06
Price (USD)/ Each
41.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
41.10
Add to cart
5,6-Dihydro-5-methyluracil, 98+%, Thermo Scientific Chemicals
Catalog numberL01996.06
Price (USD)/ Each
41.10
-
Add to cart
Chemical Identifiers
CAS696-04-8
IUPAC Name5-methyl-1,3-diazinane-2,4-dione
Molecular FormulaC5H8N2O2
InChI KeyNBAKTGXDIBVZOO-UHFFFAOYNA-N
SMILESCC1CNC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)260-268?C
FormCrystals or crystalline powder
Appearance (Color)White to pale cream
Assay (HPLC)≥98.0%
5,6-Dihydro-5-methyluracil, is used as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Dihydro-5-methyluracil, is used as an active pharmaceutical ingredient.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Luisa M.P.F. Amaral.; Piotr Szterner.; Manuel A.V. Ribeiro da Silva. Enthalpies of formation of 5,6-dihydro-5-methyluracil and 5,6-dihydro-6-methyluracil. The Journal of Chemical Thermodynamics. 2013, 64, 187-192.
  2. Paul L. Coe .; Ratnakar R. Talekar.; Richard T. Walker. The synthesis of difluoro and trifluoro analogues of pyrimidine deoxyribonucleosides: a novel approach using elemental fluorine. Journal of Fluorine Chemistry. 1994, 69, (1), 19-24.