Thermo Scientific Chemicals

5,6-Dihydro-5-methyluracil, 98+%, Thermo Scientific Chemicals

Catalog number: L01996.14
25 g, Each
Thermo Scientific Chemicals

5,6-Dihydro-5-methyluracil, 98+%, Thermo Scientific Chemicals

Catalog number: L01996.14
25 g, Each
Quantity
Catalog number: L01996.14
also known as L01996-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
696-04-8
IUPAC Name
5-methyl-1,3-diazinane-2,4-dione
Molecular Formula
C5H8N2O2
InChI Key
NBAKTGXDIBVZOO-UHFFFAOYNA-N
SMILES
CC1CNC(=O)NC1=O
Appearance (Color)
White to pale cream
Melting Point (clear melt)
260-268?C
Form
Crystals or crystalline powder
Assay (HPLC)
≥98.0%

Description

5,6-Dihydro-5-methyluracil, is used as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Dihydro-5-methyluracil, is used as an active pharmaceutical ingredient.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text