Thermo Scientific Chemicals

4,5-Dibromoveratrole, 98+%, Thermo Scientific Chemicals

Catalog number: L01999.30
250 g, Each
Thermo Scientific Chemicals

4,5-Dibromoveratrole, 98+%, Thermo Scientific Chemicals

Catalog number: L01999.30
250 g, Each
Quantity
Catalog number: L01999.30
also known as L01999-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
37895-73-1
IUPAC Name
1,2-dibromo-4,5-dimethoxybenzene
Molecular Formula
C8H8Br2O2
InChI Key
ZYCLQXMMFJREPJ-UHFFFAOYSA-N
SMILES
COC1=CC(Br)=C(Br)C=C1OC
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥98.0%
Appearance (Color)
White to pale cream to pale brown
Identification (FTIR)
Conforms

Description

4,5-Dibromoveratrole is used as chemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dibromoveratrole is used as chemical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text