Dodecanophenone, 97%, Thermo Scientific Chemicals
Dodecanophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dodecanophenone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number L02014.22
also known as L02014-22
Price (USD)/ Each
161.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
161.00
Add to cart
Dodecanophenone, 97%, Thermo Scientific Chemicals
Catalog numberL02014.22
Price (USD)/ Each
161.00
-
Add to cart
Chemical Identifiers
CAS1674-38-0
IUPAC Name1-phenyldodecan-1-one
Molecular FormulaC18H28O
InChI KeyDJNJZIFFCJTUDS-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)40.0-49.0?C
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only