Thermo Scientific Chemicals

1,2-Bis(phenylthio)ethane, 98+%, Thermo Scientific Chemicals

Catalog number: L02019.09
10 g, Each
Thermo Scientific Chemicals

1,2-Bis(phenylthio)ethane, 98+%, Thermo Scientific Chemicals

Catalog number: L02019.09
10 g, Each
Quantity
Catalog number: L02019.09
also known as L02019-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
622-20-8
IUPAC Name
{[2-(phenylsulfanyl)ethyl]sulfanyl}benzene
Molecular Formula
C14H14S2
InChI Key
MHCVYAFXPIMYRD-UHFFFAOYSA-N
SMILES
C(CSC1=CC=CC=C1)SC1=CC=CC=C1
Appearance (Color)
White to pale cream to cream to yellow
Form
Crystals or powder or crystalline powder or lumps
Assay (GC)
≥98.0%

Description

1,2-Bis(phenylthio)ethane, is used for the synthesis of the bismaleimide resin, APO-BMI, used in syntactic foams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(phenylthio)ethane, is used for the synthesis of the bismaleimide resin, APO-BMI, used in syntactic foams.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text