Thermo Scientific Chemicals

2-Ethyltoluene, 98+%, Thermo Scientific Chemicals

Catalog number: L02028.14
25 g, Each
Thermo Scientific Chemicals

2-Ethyltoluene, 98+%, Thermo Scientific Chemicals

Catalog number: L02028.14
25 g, Each
Quantity
Catalog number: L02028.14
also known as L02028-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
611-14-3
IUPAC Name
1-ethyl-2-methylbenzene
Molecular Formula
C9H12
InChI Key
HYFLWBNQFMXCPA-UHFFFAOYSA-N
SMILES
CCC1=CC=CC=C1C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
1.5020-1.5060 @ 20?C

Description

2-Ethyltoluene, is used to predict novel lead compounds for drug discovery that depends on the quality of the docking scoring function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyltoluene, is used to predict novel lead compounds for drug discovery that depends on the quality of the docking scoring function.

Solubility
Soluble in water (74.6 mg/L (25°C)).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text