Thermo Scientific Chemicals

N,N'-Diethylurea, 97%, Thermo Scientific Chemicals

Catalog number: L02079.06
5 g, Each
Thermo Scientific Chemicals

N,N'-Diethylurea, 97%, Thermo Scientific Chemicals

Catalog number: L02079.06
5 g, Each
Quantity
Catalog number: L02079.06
also known as L02079-06
Price (USD)
Quantity
-
Have Questions?

Description

N,N'-Diethylurea is used for synthesis of caffeine, theophylline, pharma chemicals, textile aids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diethylurea is used for synthesis of caffeine, theophylline, pharma chemicals, textile aids

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text