N,N'-Diethylurea, 97%, Thermo Scientific Chemicals
N,N'-Diethylurea, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N'-Diethylurea, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L02079.14
also known as L02079-14
Price (USD)/ Each
108.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
108.00
Add to cart
N,N'-Diethylurea, 97%, Thermo Scientific Chemicals
Catalog numberL02079.14
Price (USD)/ Each
108.00
-
Add to cart
Specifications
Chemical Name or MaterialN,N'-Diethylurea
CAS623-76-7
Melting Point109°C to 113°C
Recommended StorageAmbient temperatures
RTECS NumberYS9354000
View more
N,N'-Diethylurea is used for synthesis of caffeine, theophylline, pharma chemicals, textile aids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diethylurea is used for synthesis of caffeine, theophylline, pharma chemicals, textile aids

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Nabil El Tayar.; Bernard Testa.; Pierre Alain Carrupt. Polar intermolecular interactions encoded in partition coefficients: an indirect estimation of hydrogen-bond parameters of polyfunctional solutes. J. Phys. Chem. 1992, 96, (3), 1455-1459.
  2. J.P. Barbier.; R. Hugel. New complexes of cobalt(II) and nickel(II) salts with N,N'-diethylurea. Inorganica Chimica Acta. 1976, 18, 253-256.