Thermo Scientific Chemicals

4-Ethyltoluene, 97%, Thermo Scientific Chemicals

Catalog number: L02120.22
100 g, Each
Thermo Scientific Chemicals

4-Ethyltoluene, 97%, Thermo Scientific Chemicals

Catalog number: L02120.22
100 g, Each
Quantity
Catalog number: L02120.22
also known as L02120-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
622-96-8
IUPAC Name
1-ethyl-4-methylbenzene
Molecular Formula
C9H12
InChI Key
JRLPEMVDPFPYPJ-UHFFFAOYSA-N
SMILES
CCC1=CC=C(C)C=C1
Appearance (Color)
Clear, colorless
Form
Liquid
Refractive Index
1.4925-1.4975 @ 20?C
Assay (GC)
>96.0%

Description

4-Ethyltoluene is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethyltoluene is used as intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text