Thermo Scientific Chemicals

Diphenyl phosphate, 97%, Thermo Scientific Chemicals

Catalog number: L02121.22
100 g, Each
Thermo Scientific Chemicals

Diphenyl phosphate, 97%, Thermo Scientific Chemicals

Catalog number: L02121.22
100 g, Each
Quantity
Catalog number: L02121.22
also known as L02121-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
838-85-7
IUPAC Name
diphenoxyphosphinic acid
Molecular Formula
C12H11O4P
InChI Key
ASMQGLCHMVWBQR-UHFFFAOYSA-N
SMILES
OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
Appearance (Color)
White
Assay from Supplier's CofA
≥96.0% (U.S. material)
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder and/or lumps
Comment
Melting point sensitive to water content

Description

Diphenyl phosphate is used in the manufacture of phosphoric acid diesters such as triphenyl phosphate, trixylenyl phosphate, isodecyl diphenyl phosphate, cresyl diphenyl phosphate and isopropylphenyl diphenyl phosphate. It is widely used as a protective agent for hydroxyl group in organic synthesis. It finds application as an additive for paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl phosphate is used in the manufacture of phosphoric acid diesters such as triphenyl phosphate, trixylenyl phosphate, isodecyl diphenyl phosphate, cresyl diphenyl phosphate and isopropylphenyl diphenyl phosphate. It is widely used as a protective agent for hydroxyl group in organic synthesis. It finds application as an additive for paints and coatings.

Solubility
Soluble in benzene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text