Thermo Scientific Chemicals

Fluorene, 98+%, Thermo Scientific Chemicals

Catalog number: L02124.22
100 g, Each
Thermo Scientific Chemicals

Fluorene, 98+%, Thermo Scientific Chemicals

Catalog number: L02124.22
100 g, Each
Quantity
Catalog number: L02124.22
also known as L02124-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86-73-7
IUPAC Name
9H-fluorene
Molecular Formula
C13H10
InChI Key
NIHNNTQXNPWCJQ-UHFFFAOYSA-N
SMILES
C1C2=CC=CC=C2C2=CC=CC=C12
Form
Crystalline powder or powder
Assay (GC)
≥98.0%
Melting Point (clear melt)
113-117?C
Appearance (Color)
White to cream to pale brown
Identification (FTIR)
Conforms

Description

Fluorene is used in the preparation of dyes such as 2,7-diiodofluorene, 2-aminofluorene and 3,6-bis-(dimethylamino)fluorine as well as resins. It acts as a precursor to other fluorene compounds and to prepare fluorenone. It is utilized in the study of the extraction of specific semiconducting single-wall carbon nanotubes. Its polymer, polyfluorene finds application in electrically conductive and electroluminescent as well as in solar cells. In addition to this, it is used as a luminophore inorganic light-emitting diodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorene is used in the preparation of dyes such as 2,7-diiodofluorene, 2-aminofluorene and 3,6-bis-(dimethylamino)fluorine as well as resins. It acts as a precursor to other fluorene compounds and to prepare fluorenone. It is utilized in the study of the extraction of specific semiconducting single-wall carbon nanotubes. Its polymer, polyfluorene finds application in electrically conductive and electroluminescent as well as in solar cells. In addition to this, it is used as a luminophore inorganic light-emitting diodes

Solubility
Soluble in hot alcohol, ether, benzene and glacial acetic acid. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text