Thermo Scientific Chemicals

1,3,5-Trichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: L02132.18
50 g, Each
Thermo Scientific Chemicals

1,3,5-Trichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: L02132.18
50 g, Each
Quantity
Catalog number: L02132.18
also known as L02132-18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
108-70-3
IUPAC Name
1,3,5-trichlorobenzene
Molecular Formula
C6H3Cl3
InChI Key
XKEFYDZQGKAQCN-UHFFFAOYSA-N
SMILES
ClC1=CC(Cl)=CC(Cl)=C1
Identification (FTIR)
Conforms
Appearance (Color)
White to cream or pale yellow
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or lumps
Melting Point
61.5-66.5?C

Description

1,3,5-Trichlorobenzene is used in metal treatments, in industrial deodorants, in cleaners for drains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trichlorobenzene is used in metal treatments, in industrial deodorants, in cleaners for drains.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text