Thermo Scientific Chemicals

N,N'-Dimethylethylenediamine, 95%, Thermo Scientific Chemicals

Catalog number: L02204.06
5 g, Each
Thermo Scientific Chemicals

N,N'-Dimethylethylenediamine, 95%, Thermo Scientific Chemicals

Catalog number: L02204.06
5 g, Each
Quantity
Catalog number: L02204.06
also known as L02204-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-70-3
IUPAC Name
methyl[2-(methylamino)ethyl]amine
Molecular Formula
C4H12N2
InChI Key
KVKFRMCSXWQSNT-UHFFFAOYSA-N
SMILES
CNCCNC
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥94.0%
Refractive Index
1.4270-1.4330 @ 20?C

Description

N,N'-Dimethylethylenediamine is used in DNA binding effect. It is used to enhance the adsorption of carbon dioxide. It acts as a ligand and form coordination complex such as dinitrato(N,N'-dimethyl-1,2-ethanediamine)copper(II) and dichloro(1,4-bis-(diphenyl phosphino)butane)-(1,2-ethylenediamine)ruthenium(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Dimethylethylenediamine is used in DNA binding effect. It is used to enhance the adsorption of carbon dioxide. It acts as a ligand and form coordination complex such as dinitrato(N,N′-dimethyl-1,2-ethanediamine)copper(II) and dichloro(1,4-bis-(diphenyl phosphino)butane)-(1,2-ethylenediamine)ruthenium(II).

Solubility
Miscible with chloroform and dichloromethane.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text