Thermo Scientific Chemicals

Cyclopropyl phenyl ketone, 97%, Thermo Scientific Chemicals

Catalog number: L02218.22
100 g, Each
Thermo Scientific Chemicals

Cyclopropyl phenyl ketone, 97%, Thermo Scientific Chemicals

Catalog number: L02218.22
100 g, Each
Quantity
Catalog number: L02218.22
also known as L02218-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3481-02-5
IUPAC Name
cyclopropyl(phenyl)methanone
Molecular Formula
C10H10O
InChI Key
PJRHFTYXYCVOSJ-UHFFFAOYSA-N
SMILES
O=C(C1CC1)C1=CC=CC=C1
Form
Liquid
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to yellow
Identification (FTIR)
Conforms
Refractive Index
1.5510-1.5565 @ 20?C

Description

Cyclopropyl phenyl ketone is used to prepare α-cyclopropylstyrene by the Wittig reaction in dimethyl sulfoxide. It is also used as starting reagent during the (Z)-titanium enolate formation from cyclopropyl ketones via TiCl4-n-Bu4NI-induced ring opening reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropyl phenyl ketone is used to prepare α-cyclopropylstyrene by the Wittig reaction in dimethyl sulfoxide. It is also used as starting reagent during the (Z)-titanium enolate formation from cyclopropyl ketones via TiCl4-n-Bu4NI-induced ring opening reaction.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text