2-Methoxybenzylamine is used in the preparation of 6-substituted purines. It is also is used in the preparation of GABAA receptor ligands as analgesic agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methoxybenzylamine is used in the preparation of 6-substituted purines. It is also is used in the preparation of GABAA receptor ligands as analgesic agents.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
RUO – Research Use Only
Kouichi Takahashi.; Hiroto Nakano.; Reiko Fujita. A concise and practical catalytic asymmetric synthesis of (-)-CP-99,994 and (-)-L-733,061. Tetrahedron Letters. 2005, 46 (51), 8927-8930.
Kazuya Yamaguchi Dr.; Noritaka Mizuno Prof. Dr. Efficient Heterogeneous Aerobic Oxidation of Amines by a Supported Ruthenium Catalyst. Angewandte Chemie International Edition. 2003, 42(13), 1480-1483.