Diisopentylamine, 97%, Thermo Scientific Chemicals
Diisopentylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diisopentylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number L02297.14
also known as L02297-14
Price (USD)/ Each
119.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
141.00 
Save 21.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
119.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
141.00 
Save 21.35 (15%)
Add to cart
Diisopentylamine, 97%, Thermo Scientific Chemicals
Catalog numberL02297.14
Price (USD)/ Each
119.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
141.00 
Save 21.35 (15%)
-
Add to cart
Chemical Identifiers
CAS544-00-3
IUPAC Namebis(3-methylbutyl)amine
Molecular FormulaC10H23N
InChI KeySPVVMXMTSODFPU-UHFFFAOYSA-N
SMILESCC(C)CCNCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4210-1.4260 @ 20?C
Intermediate for organic synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Intermediate for organic synthesis

Solubility
Soluble in ethanol, ethyl ether and chloroform. Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Susan D. Spencer.; Cortney Bougher.; Patrick J. Heaphy.; Victor M. Murcia.; Cameron P. Gallivan.; Amber Monfette.; John D. Andersen.; Jeremy A. Cody.; Brad R. Conrad.; Christopher J. Collison. The effect of controllable thin film crystal growth on the aggregation of a novel high panchromaticity squaraine viable for organic solar cells. Solar Energy Materials and Solar Cells. 2013, 112 202-208.
  2. Christopher J. Rowlands Dr.; Lei Su Dr.; Stephen R. Elliott Prof. Rapid Prototyping of Low-Loss IR Chalcogenide-Glass Waveguides by Controlled Remelting. ChemPhysChem. 2010, 11(11), 2393-2398.