Thermo Scientific Chemicals

Tropine, 98%, Thermo Scientific Chemicals

Catalog number: L02332.09
10 g, Each
Thermo Scientific Chemicals

Tropine, 98%, Thermo Scientific Chemicals

Catalog number: L02332.09
10 g, Each
Quantity
Catalog number: L02332.09
also known as L02332-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
120-29-6
IUPAC Name
8-methyl-8-azabicyclo[3.2.1]octan-3-ol
Molecular Formula
C8H15NO
InChI Key
CYHOMWAPJJPNMW-UHFFFAOYNA-N
SMILES
CN1C2CCC1CC(O)C2
Appearance (Color)
White to pale cream
Form
Crystalline powder
Assay (GC)
> 97.5%
Water Content (Karl Fischer Titration)
< 3.0%
Assay (Non-aqueous acid-base Titration)
>97.5 to <102.5% (dry wt. basis)

Description

Tropine as an oxidative product of Tropane, used to synthesize alkaloid derivatives. Benzatropine and etybenzatropine are derivatives of tropine. It is also a building block of atropine, an anticholinergic drug prototypical of the muscarinic antagonist class.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tropine as an oxidative product of Tropane, used to synthesize alkaloid derivatives. Benzatropine and etybenzatropine are derivatives of tropine. It is also a building block of atropine, an anticholinergic drug prototypical of the muscarinic antagonist class.

Solubility
Soluble in water. (100 g/L) at 20°C.

Notes
Hygroscopic. Store at 4°C. Incomaptible with Strong acids and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text