Thermo Scientific™

Neopentyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: L02365.06
5 g, Each
Thermo Scientific™

Neopentyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: L02365.06
5 g, Each
Quantity
Catalog number: L02365.06
also known as L02365-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
753-89-9
IUPAC Name
1-chloro-2,2-dimethylpropane
Molecular Formula
C5H11Cl
InChI Key
JEKYMVBQWWZVHO-UHFFFAOYSA-N
SMILES
CC(C)(C)CCl
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Comment
Specification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)
Conforms

Description

Neopentyl chloride is a substrate used for quantum mechanics calculations on LinB to study about active site conformations and protonation states for catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Neopentyl chloride is a substrate used for quantum mechanics calculations on LinB to study about active site conformations and protonation states for catalysis.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and alkali metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text