Thermo Scientific Chemicals

Diphenyl-n-propylphosphine, 97%, Thermo Scientific Chemicals

Catalog number: L02402.06
5 g, Each
Thermo Scientific Chemicals

Diphenyl-n-propylphosphine, 97%, Thermo Scientific Chemicals

Catalog number: L02402.06
5 g, Each
Quantity
Catalog number: L02402.06
also known as L02402-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7650-84-2
IUPAC Name
diphenyl(propyl)phosphane
Molecular Formula
C15H17P
InChI Key
AAXGWYDSLJUQLN-UHFFFAOYSA-N
SMILES
CCCP(C1=CC=CC=C1)C1=CC=CC=C1
Form
Liquid
Phosphorus-31 NMR
Conforms to structure
Appearance (Color)
Clear colorless to pale yellow
Assay from Suppliers CofA
≥96.0% (GC)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text