Thermo Scientific Chemicals

Diphenylacetonitrile, 99%, Thermo Scientific Chemicals

Catalog number: L02404.36
500 g, Each
Thermo Scientific Chemicals

Diphenylacetonitrile, 99%, Thermo Scientific Chemicals

Catalog number: L02404.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L02404.36
also known as L02404-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86-29-3
IUPAC Name
2,2-diphenylacetonitrile
Molecular Formula
C14H11N
InChI Key
NEBPTMCRLHKPOB-UHFFFAOYSA-N
SMILES
N#CC(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream to yellow
Form
Crystals or powder or crystalline powder or lumps or fused solid
Assay (GC)
≥98.5%

Description

Diphenylacetonitrile undergoes anhydrous condensation with ethyl-4-bromo-butyrate. This is followed by its hydrolysis in alkaline medium that is used in the quantitative determination of 3-cyano-3,3-diphenylpropionic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylacetonitrile undergoes anhydrous condensation with ethyl-4-bromo-butyrate. This is followed by its hydrolysis in alkaline medium that is used in the quantitative determination of 3-cyano-3,3-diphenylpropionic acid.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text