Thermo Scientific Chemicals

(1-Octyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals

Catalog number: L02412.22
100 g, Each
Thermo Scientific Chemicals

(1-Octyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals

Catalog number: L02412.22
100 g, Each
Quantity
Catalog number: L02412.22
also known as L02412-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
42036-78-2
IUPAC Name
octyltriphenylphosphanium bromide
Molecular Formula
C26H32BrP
InChI Key
OBLXVLWZBMAMHE-UHFFFAOYSA-M
SMILES
[Br-].CCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to pale cream
Form
Powder or lumps
Assay (Titration ex Bromide)
≥96.0 to ≤104.0%
Melting Point (clear melt)
58.0-67.0?C

Description

(1-Octyl)triphenylphosphonium bromide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Octyl)triphenylphosphonium bromide is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text