Thermo Scientific Chemicals

2-Propanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L02431.14
25 g, Each
Thermo Scientific Chemicals

2-Propanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L02431.14
25 g, Each
Quantity
Catalog number: L02431.14
also known as L02431-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10147-37-2
IUPAC Name
propane-2-sulfonyl chloride
Molecular Formula
C3H7ClO2S
InChI Key
DRINJBFRTLBHNF-UHFFFAOYSA-N
SMILES
CC(C)S(Cl)(=O)=O
Appearance (Color)
Clear, colourless to pale yellow
Assay (Titration ex Chloride)
>96.0%
Assay (GC)
>96.0%
Refractive Index
1.4535 - 1.4585 @20?C
Form
liquid

Description

2-Propanesulfonyl chloride is used as a pharmaceutical intermediate. Reductive cleavage of a secondary alcohol to methylene has been achieved by reduction of the 2-propanesulfonyl ester with lithium triethylborohydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Propanesulfonyl chloride is used as a pharmaceutical intermediate. Reductive cleavage of a secondary alcohol to methylene has been achieved by reduction of the 2-propanesulfonyl ester with lithium triethylborohydride.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. Store under dry inert gas. It is sensitive to moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text