Bromopentamethylbenzene, 98%, Thermo Scientific Chemicals
Bromopentamethylbenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bromopentamethylbenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L02451.14
also known as L02451-14
Price (USD)/ Each
104.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
104.00
Add to cart
Bromopentamethylbenzene, 98%, Thermo Scientific Chemicals
Catalog numberL02451.14
Price (USD)/ Each
104.00
-
Add to cart
Chemical Identifiers
CAS5153-40-2
IUPAC Name1-bromo-2,3,4,5,6-pentamethylbenzene
Molecular FormulaC11H15Br
InChI KeyXPDQRULPGCFCLX-UHFFFAOYSA-N
SMILESCC1=C(C)C(C)=C(Br)C(C)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥97.0%
Melting Point (clear melt)158.5-164.5°C
Bromopentamethylbenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromopentamethylbenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Lee Irvin Smith.; James W. HornerJr. Polymethylbenzenes. XXVI. The Nitration of Bromopentamethylbenzene. J. Am. Chem. Soc. 1940, 62 (6), 1349-1354.
  2. Hitomi Suzuki. Orientation of the Products from the Nitration of Halopentamethylbenzenes. Bulletin of the Chemical Society of Japan. 1970, 43(2), 481-484.