Thermo Scientific Chemicals

2-Methylnaphthalene, 96%, Thermo Scientific Chemicals

Catalog number: L02459.22
100 g, Each
Thermo Scientific Chemicals

2-Methylnaphthalene, 96%, Thermo Scientific Chemicals

Catalog number: L02459.22
100 g, Each
Quantity
Catalog number: L02459.22
also known as L02459-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91-57-6
IUPAC Name
2-methylnaphthalene
Molecular Formula
C11H10
InChI Key
QIMMUPPBPVKWKM-UHFFFAOYSA-N
SMILES
CC1=CC=C2C=CC=CC2=C1
Appearance (Color)
White to cream to yellow to orange to pale brown
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
28.0-37.0°C
Assay (GC)
≥95.0%

Description

The main uses of methylnaphthalene are as a raw material for dyestuff dispersants and heat transfer oils, and as a solvent for agricultural chemical. It has been used as an indicator of smoke exposure in food and packaging materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The main uses of methylnaphthalene are as a raw material for dyestuff dispersants and heat transfer oils, and as a solvent for agricultural chemical. It has been used as an indicator of smoke exposure in food and packaging materials.

Solubility
Insoluble in water. Soluble in alcohol, chloroform, methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text