Thermo Scientific Chemicals

1-Pentyne, 99%, Thermo Scientific Chemicals

Catalog number: L02479.14
25 g, Each
Thermo Scientific Chemicals

1-Pentyne, 99%, Thermo Scientific Chemicals

Catalog number: L02479.14
25 g, Each
Quantity
Catalog number: L02479.14
also known as L02479-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
627-19-0
IUPAC Name
pent-1-yne
Molecular Formula
C5H8
InChI Key
IBXNCJKFFQIKKY-UHFFFAOYSA-N
SMILES
CCCC#C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.3840-1.3880 @ 20?C

Description

1-Pentyne has been used in preparation of:lithium acetylides, required for asymmetric synthesis of α,α-dibranched propargyl sulfinamides. It is also used to synthesize 7-hydroxy-10-methoxy-3H-naphtho[2.1-b]pyrans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentyne has been used in preparation of:lithium acetylides, required for asymmetric synthesis of α,α-dibranched propargyl sulfinamides. It is also used to synthesize 7-hydroxy-10-methoxy-3H-naphtho[2.1-b]pyrans.

Solubility
Soluble in water. (1.05 g/L) at 20°C.

Notes
Store at 2-8°C. Incompatible with oxidizing agents. Store away from heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text