Thermo Scientific Chemicals

2-Methylpiperidine, 98+%, Thermo Scientific Chemicals

Catalog number: L02567.AE
100 mL, Each
Thermo Scientific Chemicals

2-Methylpiperidine, 98+%, Thermo Scientific Chemicals

Catalog number: L02567.AE
100 mL, Each
Quantity
Catalog number: L02567.AE
also known as L02567-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-05-7
IUPAC Name
2-methylpiperidine
Molecular Formula
C6H13N
InChI Key
NNWUEBIEOFQMSS-UHFFFAOYNA-N
SMILES
CC1CCCCN1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay from Supplier's CofA
≥98.0% (GC)
Proton NMR
Conforms to structure

Description

2-Methylpiperidine, is used for the oxidation of secondary amines to nitrones by means of H2O2 catalyzed by Sodium tungsten oxide dihydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylpiperidine, is used for the oxidation of secondary amines to nitrones by means of H2O2 catalyzed by Sodium tungsten oxide dihydrate.

Solubility
It is easily soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text