1,2-Bis(chlorodimethylsilyl)ethane is used as Protecting Reagent for Primary Amines. It is also used in Medicine field, electronics industry and polymer material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Bis(chlorodimethylsilyl)ethane is used as Protecting Reagent for Primary Amines. It is also used in Medicine field, electronics industry and polymer material.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Incompatible with Strong oxidizing agents, Strong bases and water. Store under dry inert gas.
RUO – Research Use Only
Tetsuro Uchidaa; Yoshio Kitab; Hirofumi Maekawaa; Ikuzo Nishiguch. Mg-promoted reductive coupling of aromatic carbonyl compounds with trimethylsilyl chloride and bis(chlorodimethylsilyl) compounds.Tetrahedron.2006, 62 3103-3111.