Thermo Scientific Chemicals

Ethyl cyclohexanecarboxylate, 98+%, Thermo Scientific Chemicals

Catalog number: L02622.14
25 g, Each
Thermo Scientific Chemicals

Ethyl cyclohexanecarboxylate, 98+%, Thermo Scientific Chemicals

Catalog number: L02622.14
25 g, Each
Quantity
Catalog number: L02622.14
also known as L02622-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3289-28-9
IUPAC Name
ethyl cyclohexanecarboxylate
Molecular Formula
C9H16O2
InChI Key
JJOYCHKVKWDMEA-UHFFFAOYSA-N
SMILES
CCOC(=O)C1CCCCC1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.4395-1.4435 @ 20?C

Description

Ethyl cyclohexanecarboxylate is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cyclohexanecarboxylate is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text