1-Bromo-4-methylcyclohexane, cis + trans, 97%, Thermo Scientific Chemicals
1-Bromo-4-methylcyclohexane, cis + trans, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-4-methylcyclohexane, cis + trans, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L02640.06
also known as L02640-06
Price (USD)/ Each
81.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
81.80
Add to cart
1-Bromo-4-methylcyclohexane, cis + trans, 97%, Thermo Scientific Chemicals
Catalog numberL02640.06
Price (USD)/ Each
81.80
-
Add to cart
Chemical Identifiers
CAS6294-40-2
IUPAC Name1-bromo-4-methylcyclohexane
Molecular FormulaC7H13Br
InChI KeyRLOHLPHAOHGRNM-UHFFFAOYSA-N
SMILESCC1CCC(Br)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
1-Bromo-4-methylcyclohexane is used as intermediates in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-methylcyclohexane is used as intermediates in pharmaceutical research.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents. Incompatible with oxidizing agents.
RUO – Research Use Only