Thermo Scientific Chemicals

2,3,5-Triiodobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L02679.14
25 g, Each
Thermo Scientific Chemicals

2,3,5-Triiodobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L02679.14
25 g, Each
Quantity
Catalog number: L02679.14
also known as L02679-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-82-4
Appearance (Color)
Pale yellow to to cream to pale brown
Assay from Supplier's CofA
≥98.0%
Form
Powder
Melting Point
218-221°C

Description

2,3,5-Triiodobenzoic acid has been used in the synthesis of aromatic iodine containing vinylic monomer, 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) used in preparing radiopaque polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5-Triiodobenzoic acid has been used in the synthesis of aromatic iodine containing vinylic monomer, 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) used in preparing radiopaque polymers.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents and light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text