Thermo Scientific Chemicals

Ethyl 2-pentynoate, 97+%, Thermo Scientific Chemicals

Catalog number: L02703.14
25 g, Each
Thermo Scientific Chemicals

Ethyl 2-pentynoate, 97+%, Thermo Scientific Chemicals

Catalog number: L02703.14
25 g, Each
Quantity
Catalog number: L02703.14
also known as L02703-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55314-57-3
IUPAC Name
ethyl pent-2-ynoate
Molecular Formula
C7H10O2
InChI Key
XDPRPKSTFBPPHU-UHFFFAOYSA-N
SMILES
CCOC(=O)C#CCC
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
>97.0%
Refractive Index
1.4370-1.4420 @ 20?C

Description

Ethyl 2-pentynoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-pentynoate is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Store at 4°C. Store away from oxidizing agents. Keep away from heat. Incompatible with Oxidizing material. Acids
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text