Thermo Scientific Chemicals

3,5-Dimethyl-4-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: L02704.06
5 g, Each
Thermo Scientific Chemicals

3,5-Dimethyl-4-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: L02704.06
5 g, Each
Quantity
Catalog number: L02704.06
also known as L02704-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3095-38-3
IUPAC Name
3,5-dimethyl-4-nitrobenzoic acid
Molecular Formula
C9H9NO4
InChI Key
RBAVFNOGEPCOQI-UHFFFAOYSA-N
SMILES
CC1=CC(=CC(C)=C1[N+]([O-])=O)C(O)=O
Appearance (Color)
White to cream to pale yellow
Form
Crystals or powder or crystalline powder or fused solid
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms

Description

3,5-Dimethyl-4-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethyl-4-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text