Thermo Scientific Chemicals

4-Methylpiperidine, 98+%, Thermo Scientific Chemicals

Catalog number: L02709.AE
100 mL, Each
Thermo Scientific Chemicals

4-Methylpiperidine, 98+%, Thermo Scientific Chemicals

Catalog number: L02709.AE
100 mL, Each
Quantity
Catalog number: L02709.AE
also known as L02709-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
626-58-4
Comment
Material Sourced in the U.S. and in other countries
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.0% (non-U.S. sourced material)
Refractive Index
1.4440-1.4480 @ 20°C (non-U.S.sourced material )
Form
Liquid

Description

4-Methylpiperidine acts as a catalyst in the preparation of stilbene derivatives. It is used as an anti-corrosive compound for iron. It is also used in the synthesis of peptides to remove or deprotection of 9-fluorenylmethyloxycarbonyl (Fmoc) groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylpiperidine acts as a catalyst in the preparation of stilbene derivatives. It is used as an anti-corrosive compound for iron. It is also used in the synthesis of peptides to remove or deprotection of 9-fluorenylmethyloxycarbonyl (Fmoc) groups.

Solubility
Miscible with water and chloroform.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text