3',4'-Dimethoxyacetophenone is used as Catalytic agent; Petrochemical additive. It is also used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3′,4′-Dimethoxyacetophenone is used as Catalytic agent; Petrochemical additive. It is also used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Protect from moisture, heat. Incompatible with Strong oxidizing agents. Protect from direct light.
RUO – Research Use Only
General References:
- Henry Stephen and Charles Weizmann. CI.—Derivatives of 3 : 4-dimethoxyacetophenone and 4 : 5-dimethoxy-o-tolyl methyl ketone, and the synthesis of phenylglyoxalines containing substituents in the benzene ring.J. Chem. Soc., Trans.1914, 105 1046-1057.
- Virginia Vetere; María Belén Faraoni; Gerardo Fabián Santori; Julio César Podesta; Mónica Laura Casella; Osmar Alberto Ferretti. Study of the racemic and enantioselective hydrogenation of acetophenone and 3,4-dimethoxyacetophenone using platinum-based organotin catalysts.Catalysis Today.2005, 107 266-272.