Thermo Scientific Chemicals

8-Hydroxyquinoline sulfate monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: L02743.22
100 g, Each
Thermo Scientific Chemicals

8-Hydroxyquinoline sulfate monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: L02743.22
100 g, Each
Quantity
Catalog number: L02743.22
also known as L02743-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207386-91-2
IUPAC Name
bis(quinolin-8-ol) sulfuric acid hydrate
Molecular Formula
C18H18N2O7S
InChI Key
BNCXJZDIJIVJJO-UHFFFAOYSA-N
SMILES
O.OS(O)(=O)=O.OC1=C2N=CC=CC2=CC=C1.OC1=C2N=CC=CC2=CC=C1
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
Yellow
Assay from Suppliers CofA
≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)
≤5.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text