Thermo Scientific Chemicals

Tris(pentafluorophenyl)phosphine, 95%, Thermo Scientific Chemicals

Catalog number: L02748.03
1 g, Each
Thermo Scientific Chemicals

Tris(pentafluorophenyl)phosphine, 95%, Thermo Scientific Chemicals

Catalog number: L02748.03
1 g, Each
Quantity
Catalog number: L02748.03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1259-35-4
IUPAC Name
tris(2,3,4,5,6-pentafluorophenyl)phosphane
Molecular Formula
C18F15P
InChI Key
FQLSDFNKTNBQLC-UHFFFAOYSA-N
SMILES
FC1=C(F)C(F)=C(P(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
109.0-121.0°C
Assay (GC)
≥94.0%
Appearance (Color)
White to cream to yellow to brown

Description

Tris(pentafluorophenyl)phosphine is an additive for high voltage batteries. It acts as building blocks, coupling reagents, derivatizing reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(pentafluorophenyl)phosphine is an additive for high voltage batteries. It acts as building blocks, coupling reagents, derivatizing reagents.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances. Handle under inert gas. Incompatible with Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text