Thermo Scientific Chemicals

4-Pentyn-2-ol, 97%, Thermo Scientific Chemicals

Catalog number: L02752.06
5 g, Each
Thermo Scientific Chemicals

4-Pentyn-2-ol, 97%, Thermo Scientific Chemicals

Catalog number: L02752.06
5 g, Each
Quantity
Catalog number: L02752.06
also known as L02752-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2117-11-5
IUPAC Name
pent-4-yn-2-ol
Molecular Formula
C5H8O
InChI Key
JTHLRRZARWSHBE-UHFFFAOYNA-N
SMILES
CC(O)CC#C
Refractive Index
1.4355-1.4405 @ 20?C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%

Description

4-Pentyn-2-ol is used in Quantitative structure activity relationship (QSAR) studies pertaining to toxic levels of organic compounds in environmental situations. It is also used in the preparation of Fischer-type rhenium(III) benzoyldiazenido-2-oxacyclocarbenes. Further, it is used in the preparation of C-aryl glycosides by undergoing cycloaddition reaction with an aryl nitrile oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Pentyn-2-ol is used in Quantitative structure activity relationship (QSAR) studies pertaining to toxic levels of organic compounds in environmental situations. It is also used in the preparation of Fischer-type rhenium(III) benzoyldiazenido-2-oxacyclocarbenes. Further, it is used in the preparation of C-aryl glycosides by undergoing cycloaddition reaction with an aryl nitrile oxide.

Solubility
Miscible with chloroform and ethyl acetate.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text