Thermo Scientific Chemicals

2-Iodo-2-methylpropane, 95%, stab. with copper powder, Thermo Scientific Chemicals

Catalog number: L02786.14
25 g, Each
Thermo Scientific Chemicals

2-Iodo-2-methylpropane, 95%, stab. with copper powder, Thermo Scientific Chemicals

Catalog number: L02786.14
25 g, Each
Quantity
Catalog number: L02786.14
also known as L02786-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
558-17-8
IUPAC Name
2-iodo-2-methylpropane
Molecular Formula
C4H9I
InChI Key
ANGGPYSFTXVERY-UHFFFAOYSA-N
SMILES
CC(C)(C)I
Appearance (Color)
Clear, colourless to yellow or brown
Form
liquid
Refractive Index
1.4880-1.4940
Assay (GC)
>94.0%

Description

2-Iodo-2-methylpropane is used as a chemical reagent in organic synthesis. Further, it serves as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodo-2-methylpropane is used as a chemical reagent in organic synthesis. Further, it serves as a pharmaceutical intermediate.

Solubility
Miscible with ethanol and ether.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text