Thermo Scientific Chemicals

Hexafluoroglutaric anhydride, 97%, Thermo Scientific Chemicals

Catalog number: L02798.06
5 g, Each
Thermo Scientific Chemicals

Hexafluoroglutaric anhydride, 97%, Thermo Scientific Chemicals

Catalog number: L02798.06
5 g, Each
Quantity
Catalog number: L02798.06
also known as L02798-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
376-68-1
IUPAC Name
hexafluorooxane-2,6-dione
Molecular Formula
C5F6O3
InChI Key
IHYAGCYJVNHXCT-UHFFFAOYSA-N
SMILES
FC1(F)C(=O)OC(=O)C(F)(F)C1(F)F
Appearance (Color)
Clear colorless to pale yellow or pale pink
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.3215-1.3265 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text